Learn More
GPE Scientific™ J Young Sample Flask, Cylindrical with Fine Thread Tap
GPE Scientific™ J Young Sample Flask, Cylindrical with Fine Thread Tap is specially designed for use with vacuum manifolds and Schlenk lines.
Brand:  GPE Scientific™ JCF/50
 View more versions of this product
Product Code. 15844794
  • 89.10 € / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Solidly constructed for use with vacuum manifolds
  • Designed for use with Schlenk lines
  • Includes fine thread tap
Specifications
Specifications
50 mL
Vacuum manifolds and Schlenk lines
Fine thread tap
Sample Flask