Learn More
GPE Scientific™ J Young Sample Flask with O-Ring Tap and Cold Finger Plain Arm
GPE Scientific™ J Young Sample Flask with O-Ring Tap and Cold Finger Plain Arm is designed for use with Schlenk lines and a vacuum manifold and available in various capacities.
Brand:  GPE Scientific™ CSY/25
 View more versions of this product
Product Code. 15854744
  • 123.00 € / Each
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed with a side arm for evacuating or filling with gases
  • For connection to Schlenk lines
  • Includes O-ring tap
  • Includes cold finger plain arm
Specifications
Specifications
25 mL
O-ring tap and cold finger plain arm
Sample Flask