Learn More
GPE Scientific™ J Young Schlenk Flask, Cylindrical with Fine Thread Tap and Cone
GPE Scientific™ J Young Schlenk Flask, Cylindrical with Fine Thread Tap and Cone is specially designed for use with vacuum manifolds and Schlenk lines.
Brand:  GPE Scientific™ JCC/1000
 View more versions of this product
Product Code. 15874784
  • 151.00 € / Each
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Solidly constructed for use with vacuum manifolds
  • Designed for use with Schlenk lines
  • Includes fine thread tap and cone
Specifications
Specifications
1000 mL
Fine thread tap and cone
Size: 29
Vacuum manifolds and Schlenk lines
Schlenk Flask