Learn More
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and O-Ring Ball Joint Cup
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and O-Ring Ball Joint Cup is designed for use with Schlenk lines and a vacuum manifold and available in various capacities.
Brand:  GPE Scientific™ CSR/500
 View more versions of this product
Product Code. 15824744
  • 187.00 € / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed with a side arm for evacuating or filling with gases
  • For connection to Schlenk lines
  • Round-bottomed shape
  • Includes O-ring tap
  • Includes O-ring ball joint cup
Specifications
Specifications
500 mL
O-Ring Tap and O-Ring Ball Joint Cup
Vacuum/Schlenk Flask
Size: 29