Learn More
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and Plain Arm
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and Plain Arm is designed for use with Schlenk lines and a vacuum manifold and available in various capacities.
Brand:  GPE Scientific™ CSB/50
 View more versions of this product
Product Code. 15854734
  • 117.00 € / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed with a side arm for evacuating or filling with gases
  • For connection to Schlenk lines
  • Round-bottomed shape
  • Includes plain arm
  • Includes O-ring tap
Specifications
Specifications
50 mL
O-Ring Tap and Plain Arm
Vacuum/Schlenk Flask