Learn More
Hughes Jacketed and Insulated Tank Shower
Insulated emergency tank shower with eye/face wash unit, for use in non-hazardous hot climates. Designed to meet and exceed ANSI standards by providing tepid water, even when there is no mains water supply.
9115.00 € - 13990.00 €
Specifications
Material Stainless Steel
Product Type Tank Shower
View More Specs
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
17772348
View Documents
Hughes
HSHJ14K3SS
350 L
9115.00 €
Each
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
17762348
View Documents
Hughes
HSHJ14K1SS
1500 L
13990.00 €
Each
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
Description
Description
  • Insulated emergency tank shower with eye/face wash unit, for use in non-hazardous hot climates
  • Jacketed and insulated models maintain a steady temperature
  • Suitable for use where insufficient water pressure exists
  • Side mounted eye/face wash unit
  • Designed to meet and exceed ANSI standards by providing tepid water for over 15 minutes at a guaranteed flow of 76 liters per minute, even when there is no mains water supply
Specifications
Specifications
Stainless Steel
Tank Shower
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Hughes Jacketed and Insulated Tank Shower