missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Slide Holder
GREENER_CHOICE

Provides an inexpensive way for drying excess fluids from flat surfaces. Fisherbrand™ Slide Holder is constructed of chemical-resistant HDPE.

Brand:  Fisherbrand™ HS15981AF

30.52 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 11937984

  • 43.58 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Chemical-resistant high-density polyethylene holder

  • Convenient-to-use receptacle holds both 3 x 1 in. or 75 x 25 mm slides during evacuation and cleaning
  • Light, strong, chemical-resistant high-density polyethylene holder provides an inexpensive way for drying excess fluids from flat surfaces
  • 10° angle of slots facilitates clear viewing of desired slide
  • Beveled edges provide easy access to channels
Specifications

Specifications

40
298 x 106 x 31 mm
HDPE
Not Autoclavable
Microscope Slide
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Slide Holder > Capacity: 40 Slides

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.