Learn More
Tocris Bioscience™ Janelia Fluor™ 630b, Maleimide
Blinking fluorescent dye (Reactive group: maleimide). Application: super resolution microscopy (SRM) including SMLM
Brand:  Tocris Bioscience™ 8154/200U
Product Code. 30067471
  • 404.00 € / 200µg
Explore more special offers
Description
Description
Spontaneously Blinking Janelia Fluor™ Dyes allow facile single-molecule localization microscopy (SMLM) in cells and dense biomolecular structures, without the need for photoactivation or redox buffers. These spontaneously blinking dyes harness Janelia Fluor™ technology to delivery dyes that automatically cycle between 'off' and 'on' states with an ideal duty cycle for super resolution microscopy experiments.
Key Information: Janelia Fluor™ 630b, Maleimide is a spontaneously blinking fluorescent dye, supplied with a maleimide reactive group for conjugation (thiol reactivity).
Used for: suitable to characterize the physical properties of dense biomolecule condensates and complex macromolecular assemblies.
Application: suitable for super resolution microscopy (SRM) including single-molecule localization microscopy (SMLM).
Properties and Photophysical Data: Excitation and emission maxima (λ) are 651 nM and 666 nM, respectively.
Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.
Specifications
Specifications
N-(2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)-5,5-dimethyl-3,7-bis(3-(methylsulfonyl)azetidin-1-yl)-3'H,5H-spiro[dibenzo[b,e]siline-10,1'-isobenzofuran]-6'-carboxamide
Sulfhydyl-Reactive Janelia Fluor Dyes
0.9
200 μg
C37H40N4O8S2Si