Learn More
Tocris Bioscience™ Janelia Fluor™ 646b, Maleimide
Blinking fluorescent dye (Reactive group: maleimide). Application: super resolution microscopy (SRM) including SMLM
Brand: Tocris Bioscience™ 8158/200U
Description
Spontaneously Blinking Janelia Fluor™ Dyes allow facile single-molecule localization microscopy (SMLM) in cells and dense biomolecular structures, without the need for photoactivation or redox buffers. These spontaneously blinking dyes harness Janelia Fluor™ technology to delivery dyes that automatically cycle between 'off' and 'on' states with an ideal duty cycle for super resolution microscopy experiments.Key Information: Janelia Fluor™ 646b, Maleimide is a spontaneously blinking fluorescent dye, supplied with a maleimide reactive group for conjugation (thiol reactivity) for the labeling of protein.
Used for: suitable to characterize the physical properties of dense biomolecule condensates and complex macromolecular assemblies.
Application: suitable for super resolution microscopy (SRM) including single-molecule localization microscopy (SMLM).
Properties and Photophysical Data: derived from Janelia Fluor™ 646, Maleimide (Cat. No. 6590). Excitation and emission maxima (λ) are 662 nm and 674 nm, respectively.
Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.
Specifications
3,7-Di(azetidin-1-yl)-N-(2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)-5,5-dimethyl-3'H,5H-spiro[dibenzo[b,e]siline-10,1'-isobenzofuran]-6'-carboxamide | |
200 μg | |
C35H36N4O4Si |
2634129-70-5 | |
Fluorescent Dyes | |
0.9 |