missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ JD Honigberg™ Ice-Pak Cold Packs

Refrigerate samples for shipping with these cold packs. Cole-Parmer™ JD Honigberg™ Ice-Pak Cold Packs freeze solid in 6 to 10 hours and remain at constant -6°C for up to 72 hours.

Brand:  Cole-Parmer™ 06345-10

Product Code. 11731029

  • 152.88 € / Pack of 24

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Packs are nontoxic and won't contaminate samples
  • Use with insulated shippers and containers for best results when shipping heat-sensitive materials
  • 24 packs per case
Specifications

Specifications

24 Pk.
Refrigerate Samples For Shipping
-6°C
Cold Pack
127 x 76.2 x 25.4 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ JD Honigberg™ Ice-Pak Cold Packs > Dimensions (L x W x H): 127 x 76.2 x 25.4mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.