Learn More
Bibby Scientific™ Jenway™ Bromine Test Kit
Bibby Scientific™ Jenway™ Bromine Test Kit is a simple and convenient spectrophotometer test kit for examining water, wastewater, effluent, and environmental samples.
Marke: Bibby Scientific™ 16268-93
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- The simplest and most convenient laboratory system for measuring a broad range of parameters in water, wastewater, effluent, and environmental samples.
Spezifikation
Analyzing water, wastewater, effluent, and environmental samples. | |
100 | |
Bromide Kit |
Spectrophotometers | |
0.1 to 6.5mg/L |