man
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Cuvets and Cells
Spectroscopy Cuvets and Cells
Square or Rectangle Cuvets and Cells
Cole-Parmer™ Jenway™ Glass Cuvettes
13151734
Print
Cole-Parmer™ Jenway™ Glass Cuvettes
Capacity: Up to 14mL
Brand:
Cole-Parmer™ 99610-31
View more versions of this product
Product Code.
13151734
174.00 €
/ Each
Quantity
In Stock
Log in
to see stock.
Explore more special offers
Add to basket
This item is not returnable.
View return policy
Description
Specifications
Product Suggestions
Videos
SDS
Documents
Certificates
Special Offers
Description
Description
Specifications
Specifications
Type
Cuvette
Capacity (Metric)
14 mL
Material
Glass
Path Length
40 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers