missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jenway™ pH Electrodes
Use these electrodes for advanced applications where faster response times and greater reproducibility are required.
Brand: Jenway™ 99960-39
284.05 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
- Quickly reach equilibrium
- Significantly reduced response times
- Far greater reproducibility
- Significantly reduced long-term drift
Specifications
BNC | |
Refillable Combination with Bulb End Type | |
measurement of samples in small test tubes and Eppendorf vessels which are dissolved in Tris containing buffers | |
Glass | |
0°C to 50°C |
6 mm | |
6 dia. x 90 mm L | |
90 mm | |
0 to 14 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction