Learn More
Jenway™ pH Electrodes
Use these electrodes for advanced applications where faster response times and greater reproducibility are required.
Brand:  Jenway™ 10090-75
186.18 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
 View more versions of this product
Product Code. 11460499
  • 212.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Quickly reach equilibrium
  • Significantly reduced response times
  • Far greater reproducibility
  • Significantly reduced long-term drift
Specifications
Specifications
BNC/8 pin mini-DIN
Sealed Combination with Bulb End and Protective Cover
standard measurements of general solutions, field measurements or in education
Epoxy
0°C to 60°C
12 mm
12 dia. x 120 mm L
120 mm
0 to 14
Jenway pH meters 3505, 3510, 3520 and 370