Learn More
Jenway™ pH Electrodes
Use these electrodes for advanced applications where faster response times and greater reproducibility are required.
Brand:  Jenway™ 99960-44
 View more versions of this product
Product Code. 13107424
  • 413.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Quickly reach equilibrium
  • Significantly reduced response times
  • Far greater reproducibility
  • Significantly reduced long-term drift
Specifications
Specifications
3150/3205 waterproof connector
Sealed Combination with Bulb End and Protective Cover
measurements at low temperatures or in solutions of low ionic strength, general purpose pH measurement and accuratae Q.C. or research work
Epoxy
0°C to 80°C
12 mm
12 dia. x 120 mm L
120 mm
0 to 10