missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ JetFlex™ Genomic DNA Purification Kit

The JetFlex™ Genomic DNA Purification Kit is designed for rapid and efficient purification of genomic DNA (gDNA) from blood, body fluids (amniotic fluid, saliva, sperm, lymph), mammalian cells, tissues, plant tissues, bacteria, and yeast.

390.00 € - 1118.00 €

Specifications

Format Spin Column
Isolation Technology Tube Centrifugation
Sample Type Blood, Buccal Samples, Cells, Liquid Samples (e.g. Serum)
For Use With (Application) Real-Time PCR, PCR
High-throughput Compatibility Not High-throughput Compatible
View More Specs

Products 2
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
15637878
View Documents
Invitrogen™
A30700
100 Preps
390.00 €
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
15647878
View Documents
Invitrogen™
A30701
500 Preps
1118.00 €
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Single-tube technology allows for isolation of high quality, high molecular weight, RNA-free gDNA without the use of spin columns of a vacuum manifold.

Key benefits include:

  • Large volume processing of 2 mL of whole blood or other body fluid
  • Capable of processing:
    • 2 x 107 mammalian cells
    • 10 mg of tissue
    • 500 μL bacterial culture
    • 2 x 108 cells/mL yeast culture
  • Simple protocol that does not require columns or vacuum manifolds
Specifications

Specifications

Spin Column
Blood, Buccal Samples, Cells, Liquid Samples (e.g. Serum)
Not High-throughput Compatible
Genomic DNA Purification Kit
Genomic DNA
Tube Centrifugation
Real-Time PCR, PCR
JetFlex™
Room Temperature
Videos
SDS
Documents

Documents

Certificates

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ JetFlex™ Genomic DNA Purification Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.