Learn More
Tocris Bioscience™ JJKK 048
Potent and selective MAGL inhibitor
Brand:  Tocris Bioscience™ 5206/10
229.80 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18721809
  • 291.00 € / 10mg
Explore more special offers
Description
Description
JJKK 048 is a potent and selective monoacylglycerol lipase (MAGL) inhibitor (IC50 = 0.4 nM). Exhibits >13,000 and ∽630-fold selectivity for MAGL over FAAH and ABHD6, respectively. Increases brain levels of 2-arachidonoylglycerol (2-AG) levels in mice in vivo. Promotes analgesia in pain models.
Specifications
Specifications
>98%
JJKK 048
JJKK 048
10 mg
434.44