missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JNJ 17203212, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Reversible, competitive and potent TRPV1 antagonist (pKi values are 6.5, 7.1 and 7.3 at rat, guinea pig and human TRPV1 respectively). Inhibits capsaicin- and H+-induced channel activation (pIC50 values are 6.32 and 7.23 respectively) and exhibits antitussive and analgesic activity in vivo.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 419.32 |
Percent Purity | >99% |
Chemical Name or Material | JNJ 17203212 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'