missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JNK Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24164-1-AP-20UL
This item is not returnable.
View return policy
Description
C-Jun N-terminal Kinase (JNK) is also known as Mitogen-activated protein kinase (MAPK8). It belongs to the MAPK superfamily of stress-activated protein kinases. MAPKs are Serine-threonine protein kinases that are activated in response to a variety of extracellular stimuli, and mediate signal transduction from the cell surface to the nucleus. JNK is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. JNK pathways are activated by stress and inflammatory signals. JNK is expressed as ten different isoforms due to differential mRNA splicing. The predomit forms are JNK1 and JNK2.Specifications
JNK | |
Polyclonal | |
Unconjugated | |
MAPK10 | |
c Jun N terminal kinase 1, JNK, JNK 46, JNK1, JNK1A2, JNK21B1/2, MAP kinase 8, MAPK 8, MAPK8, PRKM8, SAPK1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26414, 26419, 26420, 5599, 5601, 5602 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P45983, P45984, P53779, Q61831, Q91Y86, Q9WTU6 | |
Mapk10, Mapk8, Mapk9 | |
JNK Fusion Protein Ag21426 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |