Learn More
JNK2 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 51153-1-AP-20UL
 View more versions of this product
Product Code. 16836425
  • 145.00 € / 20µL
Explore more special offers
Description
Description
C-Jun N-terminal Kinase (JNK) is also known as Mitogen-activated protein kinase (MAPK8). It belongs to the MAPK superfamily of stress-activated protein kinases. MAPKs are Serine-threonine protein kinases that are activated in response to a variety of extracellular stimuli, and mediate signal transduction from the cell surface to the nucleus. JNK is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. JNK pathways are activated by stress and inflammatory signals. JNK is expressed as ten different isoforms due to differential mRNA splicing. The predomit forms are JNK1 and JNK2.
Specifications
Specifications
JNK2
Polyclonal
Unconjugated
MAPK9
c Jun N terminal kinase 2, JNK 55, JNK2, JNK2A, JNK2ALPHA, JNK2B, JNK2BETA, MAP kinase 9, MAPK 9, MAPK9, p54a, p54aSAPK, PRKM9, SAPK
Rabbit
Antigen Affinity Chromatography
RUO
26420, 50658, 5601
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P45984, P49186, Q9WTU6
Mapk9
JNK2 Fusion Protein Ag1442
20 μL
Primary
Mouse, Non-human Primate, Human, Rat