missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Jumbosep™ Centrifugal Device Membrane Inserts
Jumbosep™ devices can accelerate sample processing, maximize sample recovery, add versatility, and prevent solution bypass.
Brand: Cytiva OD100C65
This item is not returnable.
View return policy
Description
- Typically concentrates 60 mL sample volumes to 5 mL in 30 minutes
- Achieve 50X concentration and > 90% recovery in just minutes
- Low protein-binding Omega™ membrane and polysulfone housing minimize losses due to non-specific binding
- Versatile Omega membrane is available in a variety of MWCOs
- Color-coded and laser etched for easy identification
- Built-in dead stop prevents spinning to dryness
- Unique sealing mechanism prevents retentate leakage and filtrate contamination
- Economical - Sample reservoir and filtrate receiver can be sanitized or autoclaved, and reused
Specifications
Clear | |
PES | |
Non-sterile | |
20 to 60 mL (Sample) |
EFA: 15.2 cm2 | |
100 kDa | |
12/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction