missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Jumbosep™ Centrifugal Device Starter Kits

Jumbosep™ Centrifugal Device Starter Kit replaces dialysis, chemical precipitation, and lyophilization.

Brand:  Cytiva (formerly Pall Lab) FD003K65

 View more versions of this product

Product Code. 17144721

  • 269.00 € / Pack of 4
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Typically concentrates 60 mL sample volumes to 5 mL in 30 minutes
  • Provides high recoveries, typically > 90%
  • For research use only
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Gray
PES
4/Pk.
20 to 60 mL (Sample)
EFA: 15.2 cm2
Non-sterile
3 kDa Starter Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers