missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Drum Safety Funnels

Prevent dangerous spills when transferring flammable liquids

Brand:  Justrite™ 08205

 View more versions of this product

Product Code. 12716055

  • 861.00 € / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket

Alert:

Drummed wastes should always be properly labeled and stored. Always segregate incompatible wastes to avoid chemical reactions or explosions.

This item is not returnable. View return policy

Description

Description

  • Large funnels have a 10.75 in. diameter (27cm) mouth and 2.6 gal. (9.8L) capacity
  • Fit 2 in. (5cm) bung on 30 and 55 gal. (114 and 208L) drums
  • Available with brass flame arrester which absorbs and dissipates heat and prevents any external ignition sources from reaching drum contents
  • Galvanized steel with durable epoxy powder-coat finish
  • Hinged, stay-open lid includes fusible link that melts at 74°C (165°F) and shuts cover to extinguish fire
  • Funnels accommodate padlock (not included)

Certifications

FM approved.

Compliance

Meet EPA and OSHA requirements.

Specifications

Specifications

FM, EPA and OSHA
Stay-open
For Flammable Liquids
32 in. Flame Arrestor, Self-closing Cover
Drum Funnel
32 L
Galvanized Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers