missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ EcoPolyblend Drumshed Accessories: Drum Ramps
Brand: Justrite™ 28679
This item is not returnable.
View return policy
Specifications
Black | |
Drum Ramp | |
1000 lb. | |
48 in. | |
11.25 in. | |
59.75 in. | |
65 lb. |
Polyethylene | |
2 to 4 drum EcoPolyBlend™ DrumShed™ | |
454 kg | |
121.92 cm | |
28.57 cm | |
151.7 cm | |
29.4 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction