missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Elite™ Smokers Cease-Fire™ Cigarette Butt Receptacles

Collect unsightly cigarette litter and reduce the risk of fire

Brand:  Justrite™ 268503

 View more versions of this product

Product Code. 12970225

  • 206.00 € / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Keep cigarette litter safely hidden from public view and enhance your property's image. Classic, contemporary upscale styling enhances or complements any commercial architectural setting.

  • No-rust polyethylene receptacle features oxygen-restricting, self-extinguishing design to safely collect burning butts
  • Stainless-steel snuffer screen offers larger snuff area for cigarettes and cigars, resulting in fewer misses
  • Limited entry design discourages unwanted trash from clogging the neck
  • Covered opening eliminates messy rainwater overflow
  • Easy cleaning and maintenance; no tools or hardware required
  • Removable, galvanized steel pail holds thousands of butts
  • For added stability and weight, add sand and/or use brick in specially designed base
  • Integrated padlock tabs accept optional padlock to discourage unauthorized access
  • For added security and greater stability in severe weather, molded-in tether eyelet accepts optional wire rope cable

Order Info

Dimensions: 30.5 dia. x 96.5Hcm (38 x 12 in. ), Capacity: 15L (4 gal.), Shipping weight: 5kg (10.4 lb.)

Compliance

ADA compliant. FM fire tested and approved.

Specifications

Specifications

15 L
4 gal.
96.5 cm
10.4 lb.
30.5 cm
12 in.
Polyethylene
FM, ADA
Black
1 Ea.
5 kg
Collecting cigarette butts
38 in.
Cigarette Butt Receptacle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.