missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Flammables Single-Door Benchtop Cabinet

Allows every lab to have a small flammables storage cabinet

Brand:  Justrite™ 890400

789.02 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


Product Code. 17828211

  • 905.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: One adjustable shelf

Description

Description

Ideal for localized storage of small quantities of flammable laboratory chemicals.

Features of Justrite Flammables Cabinets

  • Double-wall construction provides 1.5 in. of insulating air space to help prevent the spread of flames
  • High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
  • Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
  • Easy-closing, self-latching doors facilitate fire protection of the cabinet
  • Self latch engages critical three-point latching system for protection under fire conditions
  • SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
  • Exterior side panel has built-in grounding connector for easy grounding
  • Finished inside and out with durable lead-free powder paint
  • 1.5 in. (3.8cm) air space between 18G steel walls
  • 2 in. (5cm) vent openings
  • With manual closing door

Order Info

Capacity: 4 gal. (15L). Dimensions: 17L x 17W x 22 in.H (43 x 43 x 56cm). Weight: 62 lb. (28kg).

Compliance

Meet OSHA and NFPA requirements

Services

10-Year

Specifications

Specifications

NFPA, OSHA
43.2 cm
13.75 x 13.75 x 18.5 in. (34.9 x 34.9 x 47 cm)
Flammables
55.9 cm
One adjustable shelf, U-Loc™ handle with a cylinder lock and key set or an optional padlock, Haz-Alert™ reflective trilingual warning labels, leakproof 2 in. bottom sump, dual vents with flame arresters, four adjustabl
1
Epoxy, Polyester
28.12 kg
43.2 cm
Yellow
22 in.
Countertop Safety Cabinet
13.75 in.
34.9 cm
Manual-close
18.5 in.
47 cm
4 gal.
1
62 lb.
13.75 in.
34.9 cm
17 in.
Steel (CR)
17 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.