missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Sure-Grip™ EX Undercounter Corrosives/Acid Steel Safety Cabinet

Helps keep floor space free from hazards with a Sure-Grip EX Undercounter Cabinet. Suited for storing corrosives and acids away for added workplace safety.

Brand:  Justrite™ 8923021

 View more versions of this product

Product Code. 11995416

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Safely store corrosives under a workbench or lab counter for quick and easy access with these Justrite blue steel Undercounter safety cabinets for corrosives, which have the same quality safety features as the cabinets for flammables.
  • All models come with double-wall construction, dual vents, grounding wire connections, adjustable shelves, leakproof sills, three-point self-latching doors and leveling feet.
  • Self-close door(s) shuts and latches automatically when a fusible link melts at 165°F (74ºC) under fire conditions
  • Unique, concealed self-closing mechanism offers obstruction-free access to contents
  • Polyethylene trays attached to galvanized steel shelves and a separate polyethylene liner for the bottom sump resist aggressive chemicals
  • Easy-to-remove liner offers quick cleaning of drips and leaks
  • All epoxy baked-on powder-coat finish, inside and out, provides increased chemical resistance

Compliance

Self Close models meet OSHA, NFPA 30, NFPA 1 and the International Fire Code. Cabinets are FM approved.

Services

10 Years

Specifications

Specifications

FM, NFPA, OSHA, U (NFPA)
68.5 cm
18.75 x 31.75 x 27.37 in. (47.6 x 80.6 x 69.5 cm)
Flammables
88.9 cm
Double wall, dual vents, grounding wire connections, adjustable shelves, leakproof sills, three-point self latching doors and leveling feet, Polyethylene trays, separate Polyethylene liner
2
Epoxy
87 kg
88.9 cm
Blue
35 in.
Undercounter storage cabinet
18.75 in.
47.6 cm
Manual-close
27.37 in.
69.5 cm
22 gal.
1
192 lb.
31.75 in.
80.6 cm
22 in.
Steel (CR)
35 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers