missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Nonmetallic Dispensing Can
Permit easy, one-hand dispensing of hazardous liquids directly onto work surface. The nonmetallic dispensing can is made from polyethylene with a brass dispenser valve.
Brand: Justrite™ 14010
This item is not returnable.
View return policy
Description
- Ideal for use with flammables
- Leakproof, self-closing dispenser valves minimize spills, controls excess
- High-density polyethylene body
- Permits easy, one-hand dispensing of hazardous liquids directly onto work surface
Specifications
FM | |
1 qt. | |
Red | |
Polyethylene | |
4 in. |
Dispensing Can | |
0.95 L | |
For use with Flammables | |
Leakproof Brass Dispenser Valves Self-close | |
10.1 mm |