missing translation for 'onlineSavingsMsg'
Learn More

JW 480, Tocris Bioscience™

Product Code. 18035592
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective inhibitor of serine hydrolase KIAA1363 (AADACL1)

Potent and selective inhibitor of the serine hydrolase enzyme KIAA1363 (AADACL1) (IC50 = 20 nM in mouse brain). Impairs migration, invasion, survival and tumor growth of prostate cancer cell lines in vivo. Reduces monoalkylglycerol ether (MAGE) levels.

Chemical Identifiers

CAS 1354359-53-7
Molecular Formula C22H23NO2
InChI Key PCNJGBMWAZRVEA-UHFFFAOYSA-N
Synonym 2-isopropylphenyl 2-naphthalen-2-yl ethyl carbamate, 2-isopropylphenyl n-2-naphthalen-2-yl ethyl carbamate
PubChem CID 57330099
IUPAC Name (2-propan-2-ylphenyl) N-(2-naphthalen-2-ylethyl)carbamate
SMILES CC(C)C1=CC=CC=C1OC(=O)NCCC2=CC3=CC=CC=C3C=C2

Specifications

Quantity 10 mg
Formula Weight 333.42
Chemical Name or Material JW 480
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'