missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Combi Safety Storage Cabinets K-PHOENIX Vol.2-90
Safe and approved storage of aggressive and flammable hazardous substances in working areas
4056.09 € - 4282.93 €
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Depth (Metric) Exterior | 617 mm |
Color | Light Gray |
Dimensions (D x W x H) | 617 x 1197 x 1969 mm |
Material | Steel |
Alert:
For indoor use only
Product Code | Brand | Configuration | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration | Price | Quantity & Availability | |||||
15924296
|
asecos™
3033400133838 |
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated), 4 x pull-out shelf |
4056.09 €
Each |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944296
|
asecos™
3033400134256 |
3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated), 6 x pull-out shelf |
4282.93 €
Each |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Safe and approved storage of aggressive and flammable hazardous substances in working areas
Function / construction
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Two in one: 2 compartments (vertical seperated) allow the storage of flammable liquids and aggressive chemicals in one cabinet
- User-friendly and safe (folding door compartment): door open arrest system (standard), convenient and safe removal or storage of containers
- Always closed (folding door compartment): serial automatic door closing (TSA) releases the arrested door after approx. 60 seconds; acoustic and visual signal before closing
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible) / cylinder lock
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Certificates
- Fire resistance 90 minutes
- type tested in accordance with EN 14470-1
- EN 16121/16122 conformity
- GS certified
- CE compliant
Warranty
Five years
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
Light Gray | |
Steel | |
Wing, Folding, Auto-close | |
1742 mm | |
Powder-coated plain | |
450 mm |
617 mm | |
617 x 1197 x 1969 mm | |
Combi safety storage cabinets | |
1969 mm | |
2 | |
1197 mm |