missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ K-UB-90 Combination Underbench Safety Storage Cabinets



For the safe, separated and ventilated storage of slightly aggressive and flammable hazardous substances in working areas.
POA
Specifications
Certifications/Compliance | EN 14470-1, TRGS 510 |
---|---|
Color | Light Gray |
Material | Steel |
Product Type | Safety storage cabinet |
Door Style | Wing |
Product Code | Brand | Configuration | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration | Price | Quantity & Availability | |||||
15536865
|
Asecos™
|
1 x 75 kg sheet steel insert; 1 x 10 L sheet steel sump; 1 x 25 kg/11 L PP pull-out shelf with PE-sump |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15654984
![]() ![]() |
Asecos™
|
1 x 75 kg sheet steel insert; 1 x 17 L sheet steel sump; 1 x 25 kg/11 L PP pull-out shelf with PE-sump |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Separated storage compartments with vertical dividing wall
- Even and separate ventilation of each storage compartment via one central air duct (DN 75) ready for connection to a technical exhaust system
- Fits perfectly under fume cupboards and work benches to save space
- In accordance with EN 14470-1 and TRGS 510
- Uniform appearance – no motley visual appearance due to storage in one cabinet
- Can be provided with pull-out shelves, wing doors and drawers, height-adjustable shelves and bottom collecting sumps
- Fire resistance of 90 minutes
- Robust construction and longevity with body made of powder-coated sheet steel
- User-friendly and safe – door and drawer remain open in any position, interior of the cabinet completely visible, access to all containers, 93° opening of wing doors
- Safe – drawer fitted with earthing wire including clamp as standard, seal welded sump, drawers are self-closing in event of fire
- No unauthorized use – door and drawer lockable with cylinder lock and locking state indicator (red/green)
- Mobile – castors with plinth optionally available for easy moving of the cabinet
Specifications
EN 14470-1, TRGS 510 | |
Steel | |
Wing | |
300 kg |
Light Gray | |
Safety storage cabinet | |
Storage of slightly aggressive, flammable hazardous substances | |
Powder-coated |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title