Learn More
Kartell™ K120 Rubber Hoses
Kartell™ K120 Rubber Hoses are built with a cloth impression surface and are a flexible connection to various components and accessories.
Brand:  Kartell™ 0388600
 View more versions of this product
Product Code. 10214032
  • 396.00 € / 6m
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Includes cloth impression surface
  • Suitable for air, water, medium concentration acids and alcaline solutions, abrasive products, oxygenized solvents etc.
Specifications
Specifications
6 mm
18 mm
Suitable for use with air, water, medium concentration acids, alkaline solutions, abrasive products and oxygenized solvents
6 m
Rubber
6 m
-40°C
70°C