missing translation for 'onlineSavingsMsg'
Learn More

Kaempferol, 98+%, Thermo Scientific Chemicals

Product Code. 15443159
Click to view available options
Quantity:
10 mg
50 mg

Kaempferol, is used as an inhibitor of Fatty Acid Synthase, Cox-1 activity, and Topo I. It also Induces caspase-9-mediated apoptosis in a variety of cancer cell lines via downregulation of polo-like kinase 1 (PLK1) expression. Exhibits antioxidant activity and attenuates osteoclastic bone reabsorption in vitro. Also blocks EGF-induced histone H3Ser10 phosphorylation in mouse epidermal JB6 C141 cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Kaempferol, is used as an inhibitor of Fatty Acid Synthase, Cox-1 activity, and Topo I. It also Induces caspase-9-mediated apoptosis in a variety of cancer cell lines via downregulation of polo-like kinase 1 (PLK1) expression. Exhibits antioxidant activity and attenuates osteoclastic bone reabsorption in vitro. Also blocks EGF-induced histone H3Ser10 phosphorylation in mouse epidermal JB6 C141 cells.

Solubility
Soluble in DMSO or ethanol to 100mM

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Keep away fom strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 520-18-3
Molecular Formula C15H10O6
Molecular Weight (g/mol) 286.24
MDL Number MFCD00016938
InChI Key IYRMWMYZSQPJKC-UHFFFAOYSA-N
Synonym kaempferol, kaempherol, kempferol, populnetin, rhamnolutein, robigenin, trifolitin, pelargidenolon, rhamnolutin, swartziol
PubChem CID 5280863
ChEBI CHEBI:28499
SMILES OC1=CC=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C=C2O1

Specifications

Melting Point 276°C to 278°C
Color Yellow
Quantity 50 mg
Beilstein 304401
Merck Index 14,5274
Solubility Information Soluble in DMSO or ethanol to 100mM
IUPAC Name 3,5,7-trihydroxy-2-(4-hydroxyphenyl)chromen-4-one
Formula Weight 286.24
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Kaempferol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.