missing translation for 'onlineSavingsMsg'
Learn More

Karl Fischer Aqualine™ Solvent, for Karl Fischer Titration By Volumetry, Fisher Chemical™

Solvent for two-component system; Pyridine free

118.00 € - 178.00 €

Specifications

Purity or Free of Statement Pyridine free
Chemical Name or Material Aqualine™ Solvent
Color Colorless
Name Note Karl Fischer Reagent
CAS 288-32-4, 7446-09-5
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10101580
View Documents
Fisher Chemical
K/2100/15
1 L
118.00 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10264390
View Documents
Fisher Chemical
K/2100/17
2.5 L
178.00 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Solvent for two-component system; Pyridine free

  • Used in conjunction with Aqualine™ titrant for two component volumetric Karl Fischer analysis
Specifications

Specifications

Pyridine free
Colorless
288-32-4, 7446-09-5
Liquid
> 1.0
1992
Aqualine™ Solvent
Karl Fischer Reagent
Amber glass bottle
Pass Test
81734
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Karl Fischer Aqualine™ Solvent, for Karl Fischer Titration By Volumetry, Fisher Chemical™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.