missing translation for 'onlineSavingsMsg'
Learn More
Karl Fischer Reagent, for Karl Fischer Analysis, Solution ≈5mg H2O/mL Product Code.: 10538240

Karl Fischer Reagent, for Karl Fischer Analysis, Solution ≈5mg H2O/mL

Product Code. 10538240
1 L
Click to view available options
Quantity:
500 mL
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 10538240

Brand: Fisher Chemical J/4600/PB15

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

MDL Number 81734

Specifications

Color Brown
Quantity 1 L
pH 3.5 to 5.5
Packaging Plastic Coated Glass bottle
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Karl Fischer reagent
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Methyl alcohol, Pyridine, Iodine, Sulfurdioxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Karl Fischer Reagent, for Karl Fischer Analysis, Solution ≈5mg H2O/mL >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.