Learn More
Nordvlies Katrin Inclusive System Towel Dispenser
Katrin System Towel Dispenser is a quick and hygienic way to wash and dry hands with hypoallergenic tissue papers.
Brand:  Nordvlies 90045
Product Code. 15791067
  • 250.00 € / Each
Estimated Shipment: 12-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • A full range of Katrin hand drying roll tissue qualities available
  • The cover opens upwards for for easy refilling
  • Inserting a roll into the dispenser is done by simply letting the paper drop freely, pulling it until the marked line, and closing the cover
  • The fully opening cutting mechanism is easily accessible and simple to clean
  • When paper is not visible, there is an easy-to-use PUSH bar for paper feeding
  • Braille instructions are provided for visually impaired on the PUSH bar
  • Controlled consumption, minimum resistance to draw and high capacity: 420 to 800 sheets per roll depending on quality
  • The lock can be used with or without a key
  • The whole roll is always used. Almost fully used rolls automatically drop into the stub roll position when the roll has a diameter of approximately 7.5 cm
  • The empty cores can simply be pulled out side wards from the back of the dispenser and recycled
  • Main raw materials of major plastic parts and lock: ABS (Acrylonitrile butadiene styrene), PC (polycarbonate), POM (polyoxymethylene)
  • Paper consumption is easy to control, thanks to the transparent side panels
  • Katrin Inclusive dispensers are resistant to high temperatures
  • Comply with UL94 Fire Safety and Fire Protection regulations (EU)
Specifications
Specifications
Towel Dispenser
White
403 x 335 x 216 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Nordvlies Katrin Inclusive System Towel Dispenser > Tissue Dispenser; Color: White