missing translation for 'onlineSavingsMsg'
Learn More
KB-R7943 mesylate, Tocris Bioscience™ Product Code.: 18074232

KB-R7943 mesylate, Tocris Bioscience™

Product Code. 18074232
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18074232

Brand: Tocris Bioscience 1244/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Na+/Ca2+ exchange inhibitor (reverse mode)

Potent, selective inhibitor of the reverse mode of the Na+/Ca2+ exchanger (IC50 = 0.7 μM). Also inhibits the mitochondrial Ca2+ uniporter (MCU; IC50 = 5.5 μM). Does not affect Na+-dependent transport systems or ionotropic glutamate receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 182004-65-5
Molecular Formula C17H21N3O6S2
Molecular Weight (g/mol) 427.49
InChI Key WGIKEBHIKKWJLG-UHFFFAOYSA-N
Synonym kb-r7943 mesylate, 2-2-4-4-nitrobenzyloxy phenyl ethyl isothiourea mesylate, 4-4-nitrobenzyl oxy phenethyl carbamimidothioate methanesulfonate, s-4-4-nitrobenzyl oxy phenethyl isothiourea methanesulfonate, 2-2-4-4-nitrobenzyloxy phenyl ethyl isothiourea, methane sulfonate, 2-4-4-nitrophenyl methoxy phenyl ethyl sulfanylmethanimidamide; methanesulfonic acid, d0f4fkShow MoreShow Less
PubChem CID 9823846
IUPAC Name methanesulfonic acid;2-[4-[(4-nitrophenyl)methoxy]phenyl]ethyl carbamimidothioate
SMILES CS(=O)(=O)O.C1=CC(=CC=C1CCSC(=N)N)OCC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Quantity 10 mg
Formula Weight 427.49
Percent Purity >99%
Chemical Name or Material KB-R7943 mesylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
KB-R7943 mesylate, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.