missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNAB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14697-1-AP-20UL
This item is not returnable.
View return policy
Description
Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. The KCNAB1 gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily.Specifications
KCNAB1 | |
Polyclonal | |
Unconjugated | |
KCNAB1 | |
AKR6A3, hKvb3, hKvBeta3, K(+) channel subunit beta 1, KCNA1B, KCNAB1, KV BETA 1, Kvb1.3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16497, 29737, 7881 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P63143, P63144, Q14722 | |
Kcnab1 | |
KCNAB1 Fusion Protein Ag6339 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |