missing translation for 'onlineSavingsMsg'
Learn More

KCNAB3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA536970

Product Code. 15514181

  • 473.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This antibody detects endogenous protein at a molecular weight of 44 kDa. Purity is >95% by SDS-PAGE.

KCNAB3 Polyclonal antibody specifically detects KCNAB3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KCNAB3
Polyclonal
Unconjugated
KCNAB3
AKR6A9; C330022D06Rik; Ckbeta3; K(+) channel subunit beta-3; KCNA3.1B; KCNA3B; Kcnab3; Kcnab4; KV-beta.3; Kv-beta-3; mKv(beta)4; potassium channel, voltage gated shaker-related subfamily A regulatory beta subunit 3; potassium channel, voltage gated subfamily A regulatory beta subunit 3; potassium channel, voltage-dependent, beta-3 subunit; potassium channel, voltage-gated shaker-related subfamily A regulatory beta subunit 3; potassium voltage gated channel, shaker related subfamily, beta member 3; potassium voltage-gated channel subfamily A regulatory beta subunit 3; potassium voltage-gated channel, shaker-related subfamily, beta member 3; RCK beta3; Voltage-gated potassium channel subunit beta-3
Rabbit
Antigen affinity chromatography
RUO
16499, 58981, 9196
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
O43448, P97382, Q63494
KCNAB3
Synthetic peptide corresponding to amino acids 337-382 of human KV beta3
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.