Learn More
KD Scientific Stainless-Steel Syringes
Durable, electron-beam construction withstands high force, high pressure applications
1050.00 € - 1870.00 €
Specifications
Autoclavable | Autoclavable |
---|---|
For Use With | Syringe Pump |
IP Rating | None |
Material | Stainless Steel (316) |
Product Code | Brand | Diameter (Metric) Outside | Maximum Pressure | Type | Capacity (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outside | Maximum Pressure | Type | Capacity (Metric) | Price | Quantity & Availability | ||
12867362
|
KD Scientific
780801 |
12.7 mm | 7000 psi | Swagelok 1/16 in. Syringe | 2.5 mL |
1190.00 €
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
||
12887362
|
KD Scientific
780802 |
12.7 mm | 1500 psi | Swagelok 1/16 in. Syringe | 8 mL |
1145.00 €
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
||
12897362
|
KD Scientific
780803 |
22.3 mm | 1500 psi | Swagelok 1/16 in. Syringe | 20 mL |
1050.00 €
Each |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||
15164174
|
KD Scientific
780808 |
22.3 mm | 1500 psi | Swagelok 1/8 in. Syringe | 20 mL |
1050.00 €
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
||
15194174
|
KD Scientific
780812 |
22.3 mm | 1500 psi | Swagelok 1/4 in. Syringe | 20 mL |
1050.00 €
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
||
15144174
|
KD Scientific
780807 |
12.7 mm | 1500 psi | Swagelok 1/8 in. Syringe | 8 mL |
1145.00 €
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
||
11951017
|
KD Scientific
780816 |
22.3 mm | 1500 psi | Luer-Lok Syringe | 20 mL |
1050.00 €
Each |
Estimated Shipment: 17-01-2025
Log in to see stock. |
||
11986706
|
KD Scientific
780804 |
31.7 mm | 1500 psi | Swagelok 1/16 in. Syringe | 50 mL |
1050.00 €
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
||
13406038
|
KD Scientific
780809 |
31.7 mm | 1500 psi | Swagelok 1/8 in. Syringe | 50 mL |
1050.00 €
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
||
15174174
|
KD Scientific
780810 |
38.1 mm | 1500 psi | Swagelok 1/8 in. Syringe | 100 mL |
1435.00 €
Each |
In Stock
Log in to see stock. |
||
15104184
|
KD Scientific
780813 |
31.7 mm | 1500 psi | Swagelok 1/4 in. Syringe | 50 mL |
1050.00 €
Each |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||
11945596
|
KD Scientific
780805 |
38.1 mm | 1500 psi | Swagelok 1/16 in. Syringe | 100 mL |
1435.00 €
Each |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||
11971017
|
KD Scientific
780818 |
38.1 mm | 1500 psi | Luer-Lok Syringe | 100 mL |
1505.00 €
Each |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||
15114184
|
KD Scientific
780814 |
38.1 mm | 1500 psi | Swagelok 1/4 in. Syringe | 100 mL |
1435.00 €
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
||
11961017
|
KD Scientific
780817 |
31.7 mm | 1500 psi | Luer-Lok Syringe | 50 mL |
1245.00 €
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
||
17129280
|
KD Scientific
780811 |
47 mm | 1500 psi | Swagelok 1/8 in. Syringe | 200 mL |
1870.00 €
Each |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||
11905547
|
KD Scientific
780815 |
47 mm | 1500 psi | Swagelok 1/4 in. Syringe | 200 mL |
1870.00 €
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
||
Description
Eliminates hazards of glass syringe breakage.
- Compatible with most syringe pumps
- Fully autoclavable for reuse
- Adaptable to Luer Lock or Swagelok™ fittings
- Resistant to most chemicals
Specifications
Autoclavable | |
Syringe Pump | |
None | |
Stainless Steel (316) |