missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KDalert™ Lysis Buffer

The Ambion™ KDalert™ Lysis Buffer is a replacement component of the KDalert™ GAPDH Assay Kit is for the reliable measurement of GAPDH enzyme activity in cultured human, mouse, or rat cells in less than 30 minutes using a microplate fluorometer.

Brand:  Invitrogen™ AM8790G

275.37 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 10712307

  • 411.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Ambion™ KDalert™ Lysis Buffer is a replacement component of the KDalert™ GAPDH Assay Kit is for the reliable measurement of GAPDH enzyme activity in cultured human, mouse, or rat cells in less than 30 minutes using a microplate fluorometer. The buffer is supplied in one bottle of 110 mL.

Accessory Products:

The KDalert™ Kit is available (Cat. No. AM1639) and designed for use with Silencer™ GAPDH siRNAs (Cat. No. AM4605, AM4633, AM4634, AM4624, AM4632, or AM4631).

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cell Lysis Buffer
Lysis Buffer
110 mL
Liquid
Ethylene oxide (impurity) <0.01%
Ambion™, KDalert™
110 mL
KDalert™ Lysis Buffer, 110mL, Liquid, Ethylene Oxide (Impurity) <0.01% Concentration, Store at -20 to 4°C, Water Soluble, Used For RNAi
RNAi, epigenetics and non-coding RNA research, RNAi, siRNA
KDalert Lysis Buffer can be stored at -20°C or 4°C.
Dry Ice
Water Soluble
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.