Learn More
DWK Life Sciences KECK™ Assembly Set
The KECK™ assembly set serves as a connection system for a suction bottle, via which the vacuum pump can be connected.
Brand:  DWK Life Sciences 292585407
75.03 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Product Code. 12313896
  • 111.00 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Buechner funnels are cylinders with an integrated large-pored sieve plate. Without additional filter paper, only coarse filtration is possible. Buechner funnels can also be used in vacuum filtration. The applied vacuum accelerates the filtration.
  • With removable plastic hose connection (PBT), short and long screw (PP) and seals (VMQ, EPDM)
  • Suitable for filtering flasks 100 - 20000 ml
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10
Filtering, Vacuum Filtration, Safe Working Under Vacuum
9 mm
PBT, Polypropylene, VMQ, EPDM
Filtering; Vacuum filtration; Safe working under vacuum