Learn More
DWK Life Sciences KECK™ Clip Assortment, for conical joints, from stainless steel (1.4310, blank)
The KECK™ clamp is used to secure a standard ground joint connection.
Brand:  DWK Life Sciences 290301104
 View more versions of this product
Product Code. 10662183
  • 93.10 € / Pack of 10
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
KECK™ clamps are ground joint clamps used to secure standard ground joint connections in glass apparatus and are characterized by their easy handling and excellent resilience. The clamps are available in different materials (metal or polyoxymethylene).
  • Dimensionally stable with particularly good spring properties
  • Easy, safe and quick to set up
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10
Secure Glass-to-Glass Joints
NS 34
Stainless Steel (1.4310, Blank)
Secure glass-to-glass joints
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences KECK™ Clip Assortment, for conical joints, from stainless steel (1.4310, blank) > NS 34