Learn More
DWK Life Sciences™ KECK™ Clip Assortment, for spherical joints, from POM
The KECK™ clamp is used to secure a standard ground joint connection.
Brand:  DWK Life Sciences™ 290320007
41.71 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Product Code. 10283641
  • 43.90 € / Each
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
KECK™ clamps are ground joint clamps used to secure standard ground joint connections in glass apparatus and are characterized by their easy handling and excellent resilience. The clamps are available in different materials (metal or polyoxymethylene).
  • For spherical joints
  • Dimensionally stable with particularly good spring properties
  • Easy, safe and quick to set up
  • Increased safety in the laboratory as the glass surface is not scratched
  • Marking of the individual sizes for better differentiation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Ea.
2 x KS 13, 2 x KS 19, 1 x KS 29, 1 x KS 35
Secure glass-to-glass joints
POM
Secure Glass-to-Glass Joints
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences™ KECK™ Clip Assortment, for spherical joints, from POM > POM