Learn More
DWK Life Sciences KECK™ Clip for Conical Joints, Ni, KCM
KECK™ conical ground joint clamps for high temperature applications.
Brand:  DWK Life Sciences 286337508
789.72 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15901581
  • 949.00 € / Pack of 100
Estimated Shipment: 31-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
For applications at very high temperatures, the KECK™ nickel-plated spring steel ground joint clamps an important addition to KECK™ conical ground joint clips made of POM (polyoxymethylene). Compared to conventional wire clamps, they are easy and safe to attach and are inherently stable.
  • Easy and safe to put on and are dimensionally stable
  • Good dimensional stability compared to conventional wire clamps
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100
For Use at Very High Temperatures
NS 24
Nickel
For use at very high temperatures; Securing conical ground joints