Learn More
DWK Life Sciences KECK™ Clip, for conical joints, from POM
The KECK™ clamp is used to secure a standard ground joint connection.
Brand: DWK Life Sciences 1090979
43.47 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
KECK™ clamps are ground joint clamps used to secure standard ground joint connections in glass apparatus and are characterized by their easy handling and excellent resilience. The clamps are available in different materials (metal or polyoxymethylene).
- For conical joints
- Dimensionally stable with particularly good spring properties
- Easy, safe and quick to set up
- Increased safety in the laboratory as the glass surface is not scratched
- Marking of the individual sizes for better differentiation
Specifications
10 | |
Secure Glass-to-Glass Joints | |
NS 19 |
POM | |
Secure glass-to-glass joints | |
Blue |