Learn More
DWK Life Sciences KECK™ Clip for Conical Joints, POM, KC
KECK™ conical ground clamps for a safe ground joint.
Brand:  DWK Life Sciences 286336503
358.21 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15804688
  • 432.00 € / Pack of 100
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
KECK™ POM (polyoxymethylene) conical ground joint clamps are easy, safe and quick to attach and are characterized by their dimensional stability and good spring properties. In contrast to conventional wire clamps, the glass surface is not damaged when it is attached. This increases safety in the laboratory. The sizes can be easily distinguished from each other by the different colours.
  • Excellent dimensional stability and good spring properties
  • No damage to the glass surface during attachment unlike conventional wire clamps
  • Easy, safe and quick to put on and take off
  • Colour coding per size
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100
Securing of Conical Ground Joints
NS 24
POM
Securing of conical ground joints
Green