Learn More
Lenz Laborglasinstrumente™ Keck™ Conical Joint Clips - Nickel
Nickel clips for conical ground joints
Brand:  Lenz Laborglasinstrumente™ 01103924
 View more versions of this product
Product Code. 11993361
  • 119.00 € / Pack of 10
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Pack of 10
Specifications
Specifications
10
Nickel
For ground joints NS 24/29