missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences KECK™ Screw Cap for Adapter KA, with flat EPDM sealing 12 mm

Spare part for KECK™ Adapter KA.

Brand:  DWK Life Sciences 286339406

320.00 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15951581

  • 349.00 € / Pack of 100
Estimated Shipment: 04-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The screw cap as spare part for KECK™ Adapter KA is available for both thread types GL 14 and RD 14.

  • With flat gasket EPDM 12 mm
  • KECK™ Screw cap for adapter KA available for threads GL 14 and RD 14
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Red
Connection of a hose piece with a threaded cap
RD 14
Not Autoclavable
100/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.