missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences KECK™ Tubing Clamp, PBT, KT

For infinitely variable regulation of flow rates.

5.50 € - 528.00 €

Specifications

Material PBT
For Use With (Equipment) Infinitely Variable Regulation of Flow Rates
For Use With (Application) infinitely variable regulation of flow rates
View More Specs

Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15834688
View Documents
DWK Life Sciences
286338101
Fits tube OD 4.5 mm
339.00 €
Pack of 100
In Stock
to see stock.
Add to basket
 
15911581
View Documents
DWK Life Sciences
286338307
Fits tube OD 10 mm
528.00 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
15844688
View Documents
DWK Life Sciences
286338204
Fits tube OD 6 mm
506.00 €
Pack of 100
In Stock
to see stock.
Add to basket
 
10161731
View Documents
DWK Life Sciences
286338401
Fits tube OD 14 mm
5.50 €
Each
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

With just one finger, flow rates in hoses can be infinitely adjusted with KECK™ hose clamps. The clamps are available in different colours for the different hose diameters.

  • Continuous regulation of flow rates
  • Handling with one hand possible
  • Color coding of the clamps according to the different hose diameters
Specifications

Specifications

PBT
Infinitely Variable Regulation of Flow Rates
infinitely variable regulation of flow rates
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.